Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Biuret Reagent
SCI_ED

Catalog No. S25200A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25200A 500 mL
S25200 100 mL
S25200B 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S25200A
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

For protein test

For protein test

This product is for educational use - no C of A is available.

Specifications

Chemical Name or Material Biuret Reagent
Color Blue
CAS 7759-99-8, 7681-11-0, 6381-59-5, 131-73-2, 7732-18-5
Assay Percent Range <1% w/v
Packaging Solution
Quantity 500 mL
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.