Learn More
Cayman Chemical BIvalIrudIn 10mg
SDP
Catalog No. 502993944
Encompass_Preferred
A thrombin inhibitor (Kis = 2.56 and 1.84 nM for α- and ζ-thrombin, respectively); selective for α- and ζ-thrombin, lacking activity at trypsin and γ-thrombin at a >1,000-fold excess; inhibits α-thrombin-stimulated activation of the clotting factors Factor X, Factor V, and prothrombin in contact-activated plasma; reduces platelet deposition in a rat carotid endarterectomy model in a dose-dependent manner
Catalog No. 50-299-3944 Supplier Cayman Chemical Supplier No. 2303510 MG
A thrombin inhibitor (Kis = 2.56 and 1.84 nM for α- and ζ-thrombin, respectively); selective for α- and ζ-thrombin, lacking activity at trypsin and γ-thrombin at a >1,000-fold excess; inhibits α-thrombin-stimulated activation of the clotting factors Factor X, Factor V, and prothrombin in contact-activated plasma; reduces platelet deposition in a rat carotid endarterectomy model in a dose-dependent manner