Learn More
Selleck Chemical LLC Bivalirudin Trifluoroacetate 50mg 128270-60-0 D-Phe-Pro-Arg-Pro-Gly-Gly-Gly-Gly-Asn-Gly-Asp-Phe-Glu-Glu-Ile-Pro-Glu-Glu-Tyr-Leu-OH
SDP
Supplier:  Selleck Chemical LLC P100450MG
Encompass_Preferred
Bivalirudin Trifluoroacetate is a synthetic 20 residue peptide (thrombin inhibitor) which reversibly inhibits thrombin. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8040