Learn More
Medchemexpress LLC HY-10587 10mg Medchemexpress, BIX-01294 CAS:935693-62-2 Purity:>98%
SDP
Catalog No. 501968300
Encompass_Preferred
Medchemexpress, HY-10587 10mg BIX-01294 CAS:935693-62-2 BIX-01294 is an inhibitor of G9a Histone Methyltransferase with an IC50 of 1.9 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8300 Supplier Medchemexpress LLC Supplier No. HY1058710MG
Medchemexpress, HY-10587 10mg BIX-01294 CAS:935693-62-2 BIX-01294 is an inhibitor of G9a Histone Methyltransferase with an IC50 of 1.9 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.