Learn More
AdipoGen BIX 01294 (25 mg)
SDP
Supplier:  AdipoGen AGCR10051M025
Encompass_Preferred
Chemical. CAS: 935693-62-2. Formula: C28H38N6O2. MW: 490.6. Selective and cell permeable G9a histone-lysine methyltransferase (HMTase) inhibitor. Does not compete with cofactor S-adenosylmethionine. Pluripotent stem cell inducer.
Catalog No. 50-205-3807