Learn More
Enzo Life Sciences BIX 01294 (2mg). CAS: 935693-62-2
SDP
Catalog No. 502008599
Encompass_Preferred
Selective and cell permeable inhibitor of G9a histone methyltransferase. Purity: ≥95% (HPLC). Appearance: White solid. Solubility: Soluble in DMSO (5mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-8599 Supplier Enzo Life Sciences Supplier No. ALX270473M002
Selective and cell permeable inhibitor of G9a histone methyltransferase. Purity: ≥95% (HPLC). Appearance: White solid. Solubility: Soluble in DMSO (5mg/ml). Long Term Storage: -20°C.