Learn More
Medchemexpress LLC Bix-01294 (trihydrochloride) | 1392399-03-9 | MFCD16618384 | 99.7% | 600.02 g/mol | C28H41Cl3N6O2 | 250 MG
SDP
Catalog No. 5000243959
Encompass_Preferred
BIX-01294 trihydrochloride is a reversible, selective inhibitor of the histone methyltransferases G9a and GLP used for epigenetic research. It reduces H3K9 dimethylation and modulates gene expression and cell behavior in biochemical and cell-based assays; supplied as a light yellow solid with high reported purity.
  • Reversible, selective inhibitor of G9a and GLP.
  • Active in biochemical and cell assays at low micromolar concentrations.
  • Reduces H3K9me2 levels and affects gene expression.
  • High reported purity (>99%).
  • Soluble in water and DMSO at stated concentrations.
  • Powder stable at 4°C; solutions stable with frozen storage per guidance.
Catalog No. 50-002-43959 Supplier Medchemexpress LLC Supplier No. HY108239250MG
BIX-01294 trihydrochloride is a reversible, selective inhibitor of the histone methyltransferases G9a and GLP used for epigenetic research. It reduces H3K9 dimethylation and modulates gene expression and cell behavior in biochemical and cell-based assays; supplied as a light yellow solid with high reported purity.
  • Reversible, selective inhibitor of G9a and GLP.
  • Active in biochemical and cell assays at low micromolar concentrations.
  • Reduces H3K9me2 levels and affects gene expression.
  • High reported purity (>99%).
  • Soluble in water and DMSO at stated concentrations.
  • Powder stable at 4°C; solutions stable with frozen storage per guidance.