Learn More
Selleck Chemical LLC BIX-01294 trihydrochloride
SDP
Catalog No. 5000002891
Encompass_Preferred
BIX-01294 trihydrochloride is an inhibitor of G9a histone methyltransferase with IC50 of 2 7 M in a cell-free assay reduces H3K9me2 of bulk histones also weakly inhibits GLP (primarily H3K9me3) no significant activity observed at other histone methyltransferases BIX01294 induces autophagy BIX01294 also inhibits H3K36 methylation by oncoproteins NSD1 NSD2 and NSD3
Catalog No. 50-000-02891 Supplier Selleck Chemical LLC Supplier No. S800650MG
BIX-01294 trihydrochloride is an inhibitor of G9a histone methyltransferase with IC50 of 2 7 M in a cell-free assay reduces H3K9me2 of bulk histones also weakly inhibits GLP (primarily H3K9me3) no significant activity observed at other histone methyltransferases BIX01294 induces autophagy BIX01294 also inhibits H3K36 methylation by oncoproteins NSD1 NSD2 and NSD3