Learn More
Apexbio Technology LLC BIX 02189, 5mg. Cas: 1094614-85-3 MFCD: MFCD18074528. Selective MEK5 inhibitor
SDP
Supplier:  Apexbio Technology LLC A58015
Encompass_Preferred
BIX 02189 is a selective inhibitor of MEK5 with IC50 value of 1.5 nM 1.BIX 02189 belongs to the indolinone kinase inhibitor series. It selectively inhibited the catalytic activity of MEK5 but not other closely related kinases such as MEK1, MEK2, ERK2 and JNK2. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4962