Learn More
Selleck Chemical LLC BIX 02189 5mg 1094614-85-3
SDP
Catalog No. NC0360303
Encompass_Preferred
BIX02189 is a selective inhibitor of MEK5 with IC50 of 1.5 nM, also inhibits ERK5 catalytic activity with IC50 of 59 nM in cell-free assays, and does not inhibit closely related kinases MEK1, MEK2, ERK2, and JNK2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0360303 Supplier Selleck Chemical LLC Supplier No. S15315MG
BIX02189 is a selective inhibitor of MEK5 with IC50 of 1.5 nM, also inhibits ERK5 catalytic activity with IC50 of 59 nM in cell-free assays, and does not inhibit closely related kinases MEK1, MEK2, ERK2, and JNK2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.