Learn More
Apexbio Technology LLC BIX 02565, 10mg. Cas: 1311367-27-7 MFCD: MFCD25976796. RSK2 inhibitor
SDP
Supplier:  Apexbio Technology LLC B129510
Encompass_Preferred
BIX 02565 is a novel inhibitor of ribosomal S6 kinase 2 with IC50 value of 1 nM [1]. Ribosomal S6 kinase (RSK) is a Na/H exchanger (NHE) -activating factor and is important for pH maintenance during the early phase of cellular stress. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5215