Learn More
Apexbio Technology LLC BIX 02565 1311367-27-7 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B12955.1
Encompass_Preferred
BIX 02565 (CAS 1311367-27-7) is a potent inhibitor of ribosomal S6 kinase 2 (RSK2) showing an IC50 of approximately 1 nM RSK activation typically stimulates Na /H exchanger (NHE) crucial for early cellular stress responses involving intracellular pH regulation calcium handling and potential induction of cardiac hypertrophy BIX 02565 also inhibits adrenergic ( 1A 1B 1D 2A 2) imidazoline I2 LRRK2 and PKD1 receptors at varying potencies (IC50 0 052 1 820 M 16 nM and 35 nM respectively) In vivo studies in rats demonstrated dose-dependent decreases in heart rate and mean arterial pressure This compound aids research into signaling pathways relevant to cardiovascular disorders and kinase biology
Catalog No. 50-000-62304