Learn More
Cayman Chemical BIX01294 (hydrochloride hydrate), Purity: ≥98%, CAS: 1808255-64-2, 10 mg
SDP
Catalog No. 502955887
Encompass_Preferred
A selective inhibitor of G9a histone methyltransferase (IC50 = 1.7 μM); less effectively inhibits G9a-like protein (GLP; IC50 = 38 μM) and has no effect on other known histone methyltransferases
Catalog No. 50-295-5887 Supplier Cayman Chemical Supplier No. 1312410 MG
A selective inhibitor of G9a histone methyltransferase (IC50 = 1.7 μM); less effectively inhibits G9a-like protein (GLP; IC50 = 38 μM) and has no effect on other known histone methyltransferases