Learn More
Medchemexpress LLC HY-12055 5mg , BIX02188 CAS:334949-59-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY120555MG
Encompass_Preferred
HY-12055 5mg BIX02188 CAS: 334949-59-6 BIX02188 is a potent MEK5-selective inhibitor with an IC50 of 4.3 nM. BIX02188 inhibits ERK5 catalytic activity, with an IC50 of 810 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9467