Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bixin | 6983-79-5 | 10 MG
SDP

Catalog No. 5000419424
Encompass_Preferred

Bixin is an orally active carotenoid found in the seeds of Bixa orellana. It is known for inducing apoptosis in cancer cells and possessing anti-inflammatory, anti-tumor, and anti-oxidant activities. Research indicates that Bixin treatment can ameliorate cardiac dysfunction by inhibiting fibrosis, inflammation, and reactive oxygen species (ROS) generation.

  • Purity: 99.53%
  • Molecular weight: 394.50; Formula: C25H30O4
  • Appearance: Solid, brown to reddish brown
  • Source: Bixa orellana seeds
  • Induces apoptosis in cancer cells.
  • Exhibits anti-inflammatory, anti-tumor, and anti-oxidant activities.
  • Ameliorates cardiac dysfunction by inhibiting fibrosis, inflammation, and ROS generation.
  • Attenuates inflammatory response via TLR4/NF-κB signaling pathway.
  • Storage: 4°C (solid), -80°C (in solvent for 6 months), -20°C (in solvent for 1 month); protect from light.

Catalog No. 50-004-19424 Supplier Medchemexpress LLC Supplier No. HYN688410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bixin is an orally active carotenoid found in the seeds of Bixa orellana. It is known for inducing apoptosis in cancer cells and possessing anti-inflammatory, anti-tumor, and anti-oxidant activities. Research indicates that Bixin treatment can ameliorate cardiac dysfunction by inhibiting fibrosis, inflammation, and reactive oxygen species (ROS) generation.

  • Purity: 99.53%
  • Molecular weight: 394.50; Formula: C25H30O4
  • Appearance: Solid, brown to reddish brown
  • Source: Bixa orellana seeds
  • Induces apoptosis in cancer cells.
  • Exhibits anti-inflammatory, anti-tumor, and anti-oxidant activities.
  • Ameliorates cardiac dysfunction by inhibiting fibrosis, inflammation, and ROS generation.
  • Attenuates inflammatory response via TLR4/NF-κB signaling pathway.
  • Storage: 4°C (solid), -80°C (in solvent for 6 months), -20°C (in solvent for 1 month); protect from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.