Learn More
Medchemexpress LLC Bizelesin | 129655-21-6 | MFCD00892028 | 95.1% | 815.70 g·mol⁻¹ | C43H36Cl2N8O5 | 5 MG
SDP
Catalog No. 5000263393
Encompass_Preferred
Bizelesin is an AT-specific DNA alkylating agent that generates DNA interstrand crosslinks and inhibits DNA replication, used in preclinical research to evaluate anticancer activity.
  • AT-specific DNA alkylating activity.
  • Generates DNA interstrand crosslinks.
  • Effectively inhibits DNA replication.
  • Off-white to gray solid with high purity (~95%).
  • Soluble in DMSO; formulation guidance available for in vivo use.
  • Stable in solvent at -80°C for 6 months and at -20°C for 1 month when protected from light.
Catalog No. 50-002-63393 Supplier Medchemexpress LLC Supplier No. HY1113975MG
Bizelesin is an AT-specific DNA alkylating agent that generates DNA interstrand crosslinks and inhibits DNA replication, used in preclinical research to evaluate anticancer activity.
  • AT-specific DNA alkylating activity.
  • Generates DNA interstrand crosslinks.
  • Effectively inhibits DNA replication.
  • Off-white to gray solid with high purity (~95%).
  • Soluble in DMSO; formulation guidance available for in vivo use.
  • Stable in solvent at -80°C for 6 months and at -20°C for 1 month when protected from light.