Learn More
ATCC BJ; Skin Fibroblast; Human (Homo sapiens)
Catalog No. 502383310
Encompass
BJ cells are fibroblasts established from skin taken from normal foreskin from a neonatal male. Applications include toxicology research.
Catalog No. 50-238-3310 Supplier ATCC Supplier No. CRL2522
BJ cells are fibroblasts established from skin taken from normal foreskin from a neonatal male. Applications include toxicology research.