Learn More
Agilent Technologies BJ5183-AD1 ELECTROCOMP CELLS
Catalog No. 50125020
Encompass
BJ5183-AD1 Electrocompetent Cells are recombination proficient bacterial cells carrying the pAdEasy-1 plasmid that encodes the Adenovirus-5 genome (E1/E3 deleted).
Catalog No. 50-125-020 Supplier Agilent Technologies Supplier No. 200157
BJ5183-AD1 Electrocompetent Cells are recombination proficient bacterial cells carrying the pAdEasy-1 plasmid that encodes the Adenovirus-5 genome (E1/E3 deleted).