Learn More
Agilent Technologies BJ5183 ELECTROCOMPETENT CELL
Catalog No. 50125019
Encompass
BJ5183 ElectroCompetent Cells are a recombination proficient bacterial strain They supply the components necessary for the recombination event between a transfer vector and a vector containing the adenoviral genome.
Catalog No. 50-125-019 Supplier Agilent Technologies Supplier No. 200154
BJ5183 ElectroCompetent Cells are a recombination proficient bacterial strain They supply the components necessary for the recombination event between a transfer vector and a vector containing the adenoviral genome.