Learn More
Medchemexpress LLC BJE6-106 (B106) | 1564249-38-2 | 99.9% | 381.47 | C26H23NO2 | 10 MG
SDP
Catalog No. 5000226738
Encompass_Preferred
BJE6-106 (B106) is a selective protein kinase C delta (PKCδ) inhibitor supplied as a high-purity solid for research use. It induces caspase-dependent apoptosis, activates the MKK4-JNK-H2AX pathway, and shows potent activity against PKCδ with substantially lower activity versus PKCα.
  • Selective PKCδ inhibition with IC50 0.05 μM.
  • Induces caspase-dependent apoptosis and tumor-specific effects.
  • Activates the MKK4-JNK-H2AX signaling pathway.
  • High purity solid (99.85%) suitable for biochemical and cell-based assays.
  • Molecular formula C26H23NO2 and molecular weight 381.47.
Catalog No. 50-002-26738 Supplier Medchemexpress LLC Supplier No. HY11780010MG
BJE6-106 (B106) is a selective protein kinase C delta (PKCδ) inhibitor supplied as a high-purity solid for research use. It induces caspase-dependent apoptosis, activates the MKK4-JNK-H2AX pathway, and shows potent activity against PKCδ with substantially lower activity versus PKCα.
  • Selective PKCδ inhibition with IC50 0.05 μM.
  • Induces caspase-dependent apoptosis and tumor-specific effects.
  • Activates the MKK4-JNK-H2AX signaling pathway.
  • High purity solid (99.85%) suitable for biochemical and cell-based assays.
  • Molecular formula C26H23NO2 and molecular weight 381.47.