Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BJE6-106 (B106) | 1564249-38-2 | 99.9% | 381.47 | C26H23NO2 | 50 MG
SDP

Catalog No. 5000226739
Encompass_Preferred

BJE6-106 (B106) is a selective, third-generation small-molecule inhibitor of protein kinase C delta (PKCδ) used in biochemical and cell-based research to investigate PKCδ-dependent signaling and apoptosis. It is supplied as a high-purity solid and is typically dissolved in DMSO for experimental use.

  • Selective PKCδ inhibitor with IC50 = 0.05 μM and markedly reduced activity against PKCα (IC50 = 50 μM).
  • High purity solid (99.85%) suitable for biochemical assays and cell-based studies.
  • Molecular weight 381.47 and molecular formula C26H23NO2 for precise dosing and calculations.
  • Soluble in DMSO for preparation of concentrated stock solutions.
  • Available in milligram-scale quantities tailored for research applications.

Catalog No. 50-002-26739 Supplier Medchemexpress LLC Supplier No. HY11780050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BJE6-106 (B106) is a selective, third-generation small-molecule inhibitor of protein kinase C delta (PKCδ) used in biochemical and cell-based research to investigate PKCδ-dependent signaling and apoptosis. It is supplied as a high-purity solid and is typically dissolved in DMSO for experimental use.

  • Selective PKCδ inhibitor with IC50 = 0.05 μM and markedly reduced activity against PKCα (IC50 = 50 μM).
  • High purity solid (99.85%) suitable for biochemical assays and cell-based studies.
  • Molecular weight 381.47 and molecular formula C26H23NO2 for precise dosing and calculations.
  • Soluble in DMSO for preparation of concentrated stock solutions.
  • Available in milligram-scale quantities tailored for research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.