Learn More
Chemglass Life Sciences Beaker, Griffin, 100mL, Polypropylene, Graduated
SDP
Catalog No. 50872712
Encompass_Preferred
Beaker, Griffin, 100mL, Polypropylene, Graduated. Low form polypropylene beakers are chemically resistant with printed graduations that show approximated volume. Steam autoclavable at 121°C. Sold in the package quantities as listed.
Catalog No. 50-872-712 Supplier Chemglass Life Sciences Supplier No. CG110811
Add to Cart
Add to Cart
Beaker, Griffin, 100mL, Polypropylene, Graduated. Low form polypropylene beakers are chemically resistant with printed graduations that show approximated volume. Steam autoclavable at 121°C. Sold in the package quantities as listed.