Learn More
Chemglass Life Sciences Beaker, Griffin, 600mL, Polypropylene, Graduated
SDP
Catalog No. 50872716
Encompass_Preferred
Beaker, Griffin, 600mL, Polypropylene, Graduated. Low form polypropylene beakers are chemically resistant with printed graduations that show approximated volume. Steam autoclavable at 121°C. Sold in the package quantities as listed.
Catalog No. 50-872-716 Supplier Chemglass Life Sciences Supplier No. CG110815
Beaker, Griffin, 600mL, Polypropylene, Graduated. Low form polypropylene beakers are chemically resistant with printed graduations that show approximated volume. Steam autoclavable at 121°C. Sold in the package quantities as listed.