Learn More
Apexbio Technology LLC BKM120, 100mg. Cas: 944396-07-0 MFCD: MFCD18251596. Inhibitor of pan-Class I PI3K
SDP
Supplier:  Apexbio Technology LLC A3015100
Encompass_Preferred
BKM120(NVP-BKM120, Buparlisib) is a selective PI3K inhibitor of p110/// with IC50 of 52 nM/166 nM/116 nM/262 nM, respectively. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4780