Learn More
Grainger Disposable Beakers 250mL PK50
Catalog No. NC9838626
Encompass_Preferred
Disposable Polypropylene Beakers,250mL,50/pk,Resistant to many acids,organic solvents and other common reagents,Molded with a dripless lip,a rib construction that allows stacking without binding,and a flat-sitting base
Catalog No. NC9838626 Supplier Grainger Supplier No. 3UDJ6
Disposable Polypropylene Beakers,250mL,50/pk,Resistant to many acids,organic solvents and other common reagents,Molded with a dripless lip,a rib construction that allows stacking without binding,and a flat-sitting base