Learn More
TARGETMOL CHEMICALS INC BL-1020 Mesylate 1MG
SDP
Catalog No. 502877403
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. BL-1020 Mesylate (CYP-1020 Mesylate) is an orally available GABA A receptor agonist and D2 antagonist that decreases dopamine activity and enhances GABAA activity.BL-1020 Mesylate has been used in the study of schizophrenia, Alzheimer's disease, and bi-directional cognitive impairment. Purity 100%
Catalog No. 50-287-7403 Supplier TARGETMOL CHEMICALS INC Supplier No. T268341MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. BL-1020 Mesylate (CYP-1020 Mesylate) is an orally available GABA A receptor agonist and D2 antagonist that decreases dopamine activity and enhances GABAA activity.BL-1020 Mesylate has been used in the study of schizophrenia, Alzheimer's disease, and bi-directional cognitive impairment. Purity 100%