Learn More
Medchemexpress LLC HY-108596 5mg Medchemexpress, BL-1249 CAS:18200-13-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1085965MG
Encompass_Preferred
Medchemexpress, HY-108596 5mg BL-1249 CAS:18200-13-0 BL-1249 is a nonsteroidal anti-inflammatory drug (NSAID) and a potassium channel activator. BL-1249 potently activates K2P2.1 (TREK-1) and K2P10.1 (TREK-2) with EC50 values of 5.5 μM and 8.0 μM, respectively. BL-1249 extracellular application activates all TREK subfamily members but has no effect on other K2P subfamilies. BL-1249 exhibits more selective for the bladder (EC50 of 1.26 μM) than vascular tissue (EC50 of 21.0 μM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8590