Learn More
Medchemexpress LLC HY-124729 5mg Medchemexpress, BL-918 CAS:2101517-69-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1247295MG
Encompass_Preferred
Medchemexpress, HY-124729 5mg BL-918 CAS:2101517-69-3 BL-918 is an orally active UNC-51-like kinase 1 (ULK1) activator with an EC50 of 24.14 nM. BL-918 exerts its cytoprotective autophagic effect by targeting ULK complex. BL-918 has the potential for Parkinson’s disease (PD) treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0003