Learn More
Uline Ballistic Light Safety Glass
Catalog No. NC2315055
Encompass
Pair of safety glasses with tinted lenses that filter intense light to protect eyes from welding, lasers, fiber optics, UV, and IR rays while conforming to standards for high impact resistance eyewear.
Catalog No. NC2315055 Supplier Uline Supplier No. S25059
Pair of safety glasses with tinted lenses that filter intense light to protect eyes from welding, lasers, fiber optics, UV, and IR rays while conforming to standards for high impact resistance eyewear.