Learn More
Accuflow Systems Saint-Gobain BarbLock Tubing Retainer, PVDF, 1/4 in ID
Supplier:  Accuflow Systems BL135250ALPVDF
Encompass_Preferred
BarbLock ultra-secure tubing retainers provide 360° radial crimp and compression to secure flexible tubing onto barbed fittings in demanding bioprocess, medical, and laboratory fluid-transfer applications.
  • Material: PVDF
  • Tube inner diameter: 0.25 in
  • Tube outer diameter: 0.375 in
  • One-piece clamp design
  • Compatible with all flexible tubing materials
  • Constructed with USP Class VI materials
  • Packaged bag of 250
Catalog No. NC3440518