Learn More
Accuflow Systems Saint-Gobain BarbLock Retainer, PVDF, 3/8 in ID, 5/8 in OD
Supplier:  Accuflow Systems BL135375ALPVDF
Encompass_Preferred
BarbLock ultra-secure tubing retainer provides leak-resistant retention of flexible tubing on barbed fittings using a 360° radial crimp and compression design. The AL-series one-piece PVDF sleeve/collet combines simplified assembly with chemical resistance and gamma stability for use in critical fluid-transfer and single-use bioprocess applications.
  • One-piece AL-series design simplifies assembly and reduces assembly error.
  • PVDF construction is gamma stable and suitable for sterilization.
  • Provides 360° radial crimp behind the barb and 360° compression over the barb ridge for leak-resistant connections.
  • Sized for 3/8 in ID x 5/8 in OD tubing.
  • Packaged 250 per bag for bulk assembly use.
  • High heat deflection: 221-266 °F (105-130 °C) depending on test stress.
Catalog No. NC3440520