Learn More
Accuflow Systems Saint-Gobain BarbLock Tubing Retainer, PVDF, 0.500/0.750 in
Catalog No. NC3440523
Encompass_Preferred
The Saint-Gobain BarbLock AL-series one-piece PVDF tubing retainer secures flexible tubing to barbed fittings using a full 360° radial crimp and 360° compression over the barb ridge to reduce leak paths in critical fluid-transfer applications. The integrated one-piece design combines sleeve and collet to simplify assembly and reduce assembly errors.
  • Manufactured from PVDF suitable for demanding chemical and bioprocess applications.
  • Full 360° radial crimp behind the barb for secure retention.
  • 360° compression directly over the barb ridge to minimize leak paths.
  • Compatible with a wide range of flexible tubing materials.
  • Eliminates need for solvent bonding, ultraviolet bonding, and adhesives.
  • Resins meet USP <88> Class VI and ISO 10993-5 where specified.
  • Quick and easy to assemble, gamma stable.
Catalog No. NC3440523 Supplier Accuflow Systems Supplier No. BL135500ALPVDF
The Saint-Gobain BarbLock AL-series one-piece PVDF tubing retainer secures flexible tubing to barbed fittings using a full 360° radial crimp and 360° compression over the barb ridge to reduce leak paths in critical fluid-transfer applications. The integrated one-piece design combines sleeve and collet to simplify assembly and reduce assembly errors.
  • Manufactured from PVDF suitable for demanding chemical and bioprocess applications.
  • Full 360° radial crimp behind the barb for secure retention.
  • 360° compression directly over the barb ridge to minimize leak paths.
  • Compatible with a wide range of flexible tubing materials.
  • Eliminates need for solvent bonding, ultraviolet bonding, and adhesives.
  • Resins meet USP <88> Class VI and ISO 10993-5 where specified.
  • Quick and easy to assemble, gamma stable.