Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuflow Systems Saint-Gobain BarbLock One-Piece Retainer, AL Series, PVDF, 3/4 in ID

Supplier:  Accuflow Systems BL135750ALPVDF

Encompass_Preferred

Saint-Gobain BarbLock one-piece AL series PVDF retainers provide an ultra-secure, adhesive-free method for securing flexible tubing to barbed fittings. The PVDF construction supports higher-temperature and chemically demanding environments and installs quickly for disposable or permanent assemblies in bioprocess and fluid-transfer systems.

  • Material: PVDF
  • Series: AL one-piece retainer
  • Fits tubing inner diameter: 19 mm (3/4 in)
  • Packaging: bag of 25
  • Key features: 360° radial crimp behind the barb, 360° compression over the barb, no adhesives or solvents required
  • Applications: biopharmaceutical transport, disposable assemblies, dialysis and IV equipment

Catalog No. NC3440526


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.