Learn More
Agilent Technologies BL21-CODONPLUS(DE3)RILC.CELLS
Catalog No. 50125351
Encompass
BL21-CodonPlus -RIL and -RP Competent Cells Correct codon bias, dramatically improves expression when codon bias is a problem, includes RIL strain for AT-rich genomes, RP strain for GC-rich genomes
Catalog No. 50-125-351 Supplier Agilent Technologies Supplier No. 230245
BL21-CodonPlus -RIL and -RP Competent Cells Correct codon bias, dramatically improves expression when codon bias is a problem, includes RIL strain for AT-rich genomes, RP strain for GC-rich genomes