Learn More
Gold Biotechnology Inc BL21 Competent E coli Cells
SDP
Catalog No. 5000562242
Encompass_Preferred
GoldBio s BL21 Chemically Competent E coli cells are a good universal strain of cell that is gives great transformation for protein expression and induction and is especially useful for non-T7 vectors protein expression systems BL21 chemically competent cells feature a widely used host background and are deficient in both lon (1) and ompT proteases In addition BL21 Chemically Competent E coli cells are resistant to phage T1 ( fhuA2 )
Catalog No. 50-005-62242 Supplier Gold Biotechnology Inc Supplier No. CC10215X50
GoldBio s BL21 Chemically Competent E coli cells are a good universal strain of cell that is gives great transformation for protein expression and induction and is especially useful for non-T7 vectors protein expression systems BL21 chemically competent cells feature a widely used host background and are deficient in both lon (1) and ompT proteases In addition BL21 Chemically Competent E coli cells are resistant to phage T1 ( fhuA2 )