Learn More
New England Biolabs, Inc. BL21(DE3) Competent E. coli – 6 x 0.2 ml/tube
SDP
Catalog No. NC0077830
Encompass
Chemically competent E. coli cells suitable for transformation and protein expression. BL21 (DE3) Competent E. coli is a widely used T7 expression E. coli strain.
Catalog No. NC0077830 Supplier New England Biolabs, Inc. Supplier No. C2527I
Chemically competent E. coli cells suitable for transformation and protein expression. BL21 (DE3) Competent E. coli is a widely used T7 expression E. coli strain.