Learn More
Gold Biotechnology Inc BL21 DE3 Comp E coli Cells
SDP
Catalog No. 5000562323
Encompass_Preferred
GoldBio s BL21 (DE3) Chemically Competent E coli cells are high efficiency cells suitable for a wide variety of applications such as transformation and routine protein expression BL21 (DE3) chemically competent cells feature a widely used host background a T7 expression strain and are deficient in both lon (1) and ompT proteases In addition BL21 (DE3) cells are resistant to phage T1 ( fhuA2 ) and are B strain
Catalog No. 50-005-62323 Supplier Gold Biotechnology Inc Supplier No. CC10315X50
GoldBio s BL21 (DE3) Chemically Competent E coli cells are high efficiency cells suitable for a wide variety of applications such as transformation and routine protein expression BL21 (DE3) chemically competent cells feature a widely used host background a T7 expression strain and are deficient in both lon (1) and ompT proteases In addition BL21 (DE3) cells are resistant to phage T1 ( fhuA2 ) and are B strain