Learn More
Gold Biotechnology Inc BL21 (DE3) Chemically Competent E. coli Cells 5x50 μl
SDP
Catalog No. NC2942837
Encompass_Preferred
GoldBio’s BL21 (DE3) Chemically Competent E. coli cells are suitable for transformation and routine protein expression. BL21 (DE3) chemically competent cells feature a widely used host background, a T7 expression strain, and are deficient in both Ion (1) and ompT proteases. In addition, BL21 (DE3) cells are resistant to phage T1 (fhuA2) and are B strain.
Catalog No. NC2942837 Supplier Gold Biotechnology Inc Supplier No. CC1035X50
GoldBio’s BL21 (DE3) Chemically Competent E. coli cells are suitable for transformation and routine protein expression. BL21 (DE3) chemically competent cells feature a widely used host background, a T7 expression strain, and are deficient in both Ion (1) and ompT proteases. In addition, BL21 (DE3) cells are resistant to phage T1 (fhuA2) and are B strain.