Learn More
Agilent Technologies BL21(DE3) COMPETENT CELLS
Catalog No. 50125011
Encompass
BL21(DE3) Competent Cells shows high-level protein expression, encodes T7 RNA polymerase under the control of the lacUV5 promoter. Used for non-toxic proteins.
Catalog No. 50-125-011 Supplier Agilent Technologies Supplier No. 200131
BL21(DE3) Competent Cells shows high-level protein expression, encodes T7 RNA polymerase under the control of the lacUV5 promoter. Used for non-toxic proteins.