Learn More
AMID BIOSCIENCES Amid Biosciences, BL21(DE3) Delta SerC Chemically Competent E. coli Cells, 10 x 0.1 mL
SDP
Catalog No. NC4043466
Encompass_Preferred
BL21(DE3) Delta SerC chemically competent E. coli cells are supplied in a 10 x 0.1 mL format for bacterial expression workflows involving protein phosphorylation related studies. The strain provides a specialized host background for researchers optimizing systems that require altered serine pathway context or phosphoserine incorporation strategies. The chemically competent format supports routine plasmid transformation and expression screening. Technical specifications are included below.- Quantity - 10 x 0.1 mL- Organism - E. coli- Storage - -80 C- Cell Strain / Host - BL21(DE3) delta serC- Transformation Efficiency - 1.00E+6- Cell Format - 10 x 0.1 mL chemically competent cellsThis product is for research use only.
Catalog No. NC4043466 Supplier AMID BIOSCIENCES Supplier No. BLPC201
BL21(DE3) Delta SerC chemically competent E. coli cells are supplied in a 10 x 0.1 mL format for bacterial expression workflows involving protein phosphorylation related studies. The strain provides a specialized host background for researchers optimizing systems that require altered serine pathway context or phosphoserine incorporation strategies. The chemically competent format supports routine plasmid transformation and expression screening. Technical specifications are included below.- Quantity - 10 x 0.1 mL- Organism - E. coli- Storage - -80 C- Cell Strain / Host - BL21(DE3) delta serC- Transformation Efficiency - 1.00E+6- Cell Format - 10 x 0.1 mL chemically competent cellsThis product is for research use only.