Learn More
AMID BIOSCIENCES Amid Biosciences, BL21(DE3) Delta SerC Delta YcdX Chemically Competent Cells, 10 x 0.1 mL
SDP
Catalog No. NC4043464
Encompass_Preferred
BL21(DE3) Delta SerC Delta YcdX chemically competent cells are supplied in a 10 x 0.1 mL format for specialized bacterial expression workflows. Applications include recombinant protein expression and phosphoserine incorporation workflows requiring engineered BL21(DE3) host background. Technical specifications are included below.- Quantity - 10 x 0.1 mL- Organism - E. coli- Storage - -80 C- Cell Strain / Host - BL21(DE3) delta serC delta ycdX- Transformation Efficiency - 1.00E+6- Cell Format - 10 x 0.1 mL chemically competent cellsThis product is for research use only.
Catalog No. NC4043464 Supplier AMID BIOSCIENCES Supplier No. BLPCY201
BL21(DE3) Delta SerC Delta YcdX chemically competent cells are supplied in a 10 x 0.1 mL format for specialized bacterial expression workflows. Applications include recombinant protein expression and phosphoserine incorporation workflows requiring engineered BL21(DE3) host background. Technical specifications are included below.- Quantity - 10 x 0.1 mL- Organism - E. coli- Storage - -80 C- Cell Strain / Host - BL21(DE3) delta serC delta ycdX- Transformation Efficiency - 1.00E+6- Cell Format - 10 x 0.1 mL chemically competent cellsThis product is for research use only.