Learn More
Gold Biotechnology Inc BL21 DE3 Electro E coli Cells
SDP
Catalog No. 5000562232
Encompass_Preferred
GoldBio s BL21 (DE3) Electrocompetent E coli cells are suitable for high efficiency transformation and routine protein expression Increased cloning efficiencies versus typical BL21 cells makes the BL21 (DE3) Electrocompetent E coli cells ideal for construction of complex expression libraries and feature 1 x 10 8 cfu/ g efficiency with electroporation BL21 (DE3) Electrocompetent E coli cells have a widely used background feature a T7 expression strain are deficient in both lon (1) and ompT proteases These cells are resistant to phage T1( fhuA2 ) and are B strain
Catalog No. 50-005-62232 Supplier Gold Biotechnology Inc Supplier No. CC2045X50
GoldBio s BL21 (DE3) Electrocompetent E coli cells are suitable for high efficiency transformation and routine protein expression Increased cloning efficiencies versus typical BL21 cells makes the BL21 (DE3) Electrocompetent E coli cells ideal for construction of complex expression libraries and feature 1 x 10 8 cfu/ g efficiency with electroporation BL21 (DE3) Electrocompetent E coli cells have a widely used background feature a T7 expression strain are deficient in both lon (1) and ompT proteases These cells are resistant to phage T1( fhuA2 ) and are B strain