Learn More
AMID BIOSCIENCES Amid Biosciences, BL21(DE3) Electrocompetent E. coli Cells, Recombinant Protein Expression, 10 x 0.05 mL
SDP
Catalog No. NC4043485
Encompass_Preferred
BL21(DE3) electrocompetent E. coli cells are supplied in a 10 x 0.05 mL format for high level recombinant protein expression. BL21(DE3) is a widely used T7 RNA polymerase expression host for plasmids under T7 promoter control. Applications include high-level recombinant protein expression from T7 promoter based expression vectors. Technical specifications are included below.- Quantity - 10 x 0.05 mL- Organism - E. coli- Storage - -80 C- Cell Strain / Host - BL21(DE3)- Cell Format - 10 x 0.05 mL electrocompetent cells- Transformation Efficiency - (1.00 - 2.00)E+9This product is for research use only.
Catalog No. NC4043485 Supplier AMID BIOSCIENCES Supplier No. EBL21201
BL21(DE3) electrocompetent E. coli cells are supplied in a 10 x 0.05 mL format for high level recombinant protein expression. BL21(DE3) is a widely used T7 RNA polymerase expression host for plasmids under T7 promoter control. Applications include high-level recombinant protein expression from T7 promoter based expression vectors. Technical specifications are included below.- Quantity - 10 x 0.05 mL- Organism - E. coli- Storage - -80 C- Cell Strain / Host - BL21(DE3)- Cell Format - 10 x 0.05 mL electrocompetent cells- Transformation Efficiency - (1.00 - 2.00)E+9This product is for research use only.