Learn More
AMID BIOSCIENCES Amid Biosciences, BL21(DE3) pLPP Co-Expression System, Lambda Phosphatase, 10 x 0.1 mL
SDP
Catalog No. NC4043461
Encompass_Preferred
BL21(DE3) pLPP co-expression system is supplied as competent cells in a 10 x 0.1 mL format for production of unphosphorylated recombinant proteins. Applications include co-expression of lambda protein phosphatase for production of unphosphorylated recombinant proteins and phosphoprotein control workflows. Technical specifications are included below.- Quantity - 10 x 0.1 mL- Organism - E. coli- Storage - -80 C- Cell Strain / Host - BL21(DE3) pLPP- Transformation Efficiency - 1.00E+6- Cell Format - 10 x 0.1 mL chemically competent cellsThis product is for research use only.
Catalog No. NC4043461 Supplier AMID BIOSCIENCES Supplier No. BLPP201
BL21(DE3) pLPP co-expression system is supplied as competent cells in a 10 x 0.1 mL format for production of unphosphorylated recombinant proteins. Applications include co-expression of lambda protein phosphatase for production of unphosphorylated recombinant proteins and phosphoprotein control workflows. Technical specifications are included below.- Quantity - 10 x 0.1 mL- Organism - E. coli- Storage - -80 C- Cell Strain / Host - BL21(DE3) pLPP- Transformation Efficiency - 1.00E+6- Cell Format - 10 x 0.1 mL chemically competent cellsThis product is for research use only.