Learn More
Agilent Technologies BL21(DE3) PLYSS COMP CELLS
Catalog No. 50125012
Encompass
BL21(DE3) pLysS Competent Cell show high-level protein expression, codes for T7 lysozyme and a T7 RNA polymerase inhibitor,provide tighter control of protein expression for expression of toxic proteins,can also be used for non-toxic protien
Catalog No. 50-125-012 Supplier Agilent Technologies Supplier No. 200132
BL21(DE3) pLysS Competent Cell show high-level protein expression, codes for T7 lysozyme and a T7 RNA polymerase inhibitor,provide tighter control of protein expression for expression of toxic proteins,can also be used for non-toxic protien