Learn More
Agilent Technologies BL21-GOLD COMPETENT CELLS
Catalog No. 50125347
Encompass
BL21-Gold Competent Cells 10 x 0.1mL For extremely toxic proteins use with lambda CE6 under the control of T7 RNA polymerase or use with non-T7 polymerase-based system. Efficiency is= 1 x 108 cfu/microgram of supercoiled DNA
Catalog No. 50-125-347 Supplier Agilent Technologies Supplier No. 230130
BL21-Gold Competent Cells 10 x 0.1mL For extremely toxic proteins use with lambda CE6 under the control of T7 RNA polymerase or use with non-T7 polymerase-based system. Efficiency is= 1 x 108 cfu/microgram of supercoiled DNA