Learn More
Agilent Technologies BL21-GOLD (DE3) COMP CELLS
Catalog No. 50125348
Encompass
BL21-Gold (DE3) Competent Cells Provide high-level protein expression and Allow direct cloning of many expression constructs. Hte phenotype increases transformation efficiency to 1 x 108 of supercoiled DNA. Used for extremely toxic protei
Catalog No. 50-125-348 Supplier Agilent Technologies Supplier No. 230132
BL21-Gold (DE3) Competent Cells Provide high-level protein expression and Allow direct cloning of many expression constructs. Hte phenotype increases transformation efficiency to 1 x 108 of supercoiled DNA. Used for extremely toxic protei