Learn More
AMID BIOSCIENCES Amid Biosciences, BL21(DE3) pBirA Chemically Competent E. coli Cells, In Vivo Biotinylation, 10 x 0.1 mL
SDP
Catalog No. NC2302277
Encompass
BL21(DE3) pBirA chemically competent E. coli cells are supplied in a 10 x 0.1 mL format for in vivo biotinylation workflows. The host expresses BirA biotin ligase to support biotinylation of proteins carrying an AviTag or compatible biotin acceptor peptide during bacterial expression. Applications include in vivo biotinylation of AviTag or biotin acceptor peptide tagged proteins during recombinant expression. Technical specifications are included below.- Quantity - 10 x 0.1 mL (20 reactions)- Organism - E. coli- Storage - -80 C- Cell Strain / Host - BL21(DE3) pBirA- Transformation Efficiency - 1.00E+6- Cell Format - 10 x 0.1 mL chemically competent cellsThis product is for research use only.
Catalog No. NC2302277 Supplier AMID BIOSCIENCES Supplier No. BLB201
BL21(DE3) pBirA chemically competent E. coli cells are supplied in a 10 x 0.1 mL format for in vivo biotinylation workflows. The host expresses BirA biotin ligase to support biotinylation of proteins carrying an AviTag or compatible biotin acceptor peptide during bacterial expression. Applications include in vivo biotinylation of AviTag or biotin acceptor peptide tagged proteins during recombinant expression. Technical specifications are included below.- Quantity - 10 x 0.1 mL (20 reactions)- Organism - E. coli- Storage - -80 C- Cell Strain / Host - BL21(DE3) pBirA- Transformation Efficiency - 1.00E+6- Cell Format - 10 x 0.1 mL chemically competent cellsThis product is for research use only.