Learn More
INTACT GENOMICS INC BL21(DE3) Chemically Competent Cells 6x200µl
Catalog No. 5000083052
Encompass
Intact Genomics BL21(DE3) Chemically Competent E. coli Cells are suitable for transformation and routine protein expression.BL21(DE3) Chemically Competent Cells have the following features: Widely used host background T7 Expression Strain Deficient in both lon (1) and ompT proteases Resistant to phage T1 (fhuA2) B StrainProduct Includes and Storage:BL21(DE3) Competent Cells: -80 ºCpUC19 Control DNA: -20 ºCRecovery Medium: 4 ºC
Catalog No. 50-000-83052 Supplier INTACT GENOMICS INC Supplier No. 105424
Intact Genomics BL21(DE3) Chemically Competent E. coli Cells are suitable for transformation and routine protein expression.BL21(DE3) Chemically Competent Cells have the following features: Widely used host background T7 Expression Strain Deficient in both lon (1) and ompT proteases Resistant to phage T1 (fhuA2) B StrainProduct Includes and Storage:BL21(DE3) Competent Cells: -80 ºCpUC19 Control DNA: -20 ºCRecovery Medium: 4 ºC