Learn More
Gold Biotechnology Inc BL21 (DE3) Electrocompetent E. coli Cells 5x50 μl
SDP
Catalog No. NC2942860
Encompass_Preferred
GoldBio’s BL21 (DE3) Electrocompetent E. coli cells are suitable for high efficiency transformation and routine protein expression. Increased cloning efficiencies versus typical BL21 cells makes the BL21 (DE3) Electrocompetent E. coli cells ideal for construction of complex expression libraries and feature ≥1 x 108 cfu/µg efficiency with electroporation. BL21 (DE3) Electrocompetent E. coli cells have a widely used background, feature a T7 expression strain, are deficient in both lon (1) and ompT proteases. These cells are resistant to phage T1(fhuA2) and are B strain.
Catalog No. NC2942860 Supplier Gold Biotechnology Inc Supplier No. CC2045X50C
GoldBio’s BL21 (DE3) Electrocompetent E. coli cells are suitable for high efficiency transformation and routine protein expression. Increased cloning efficiencies versus typical BL21 cells makes the BL21 (DE3) Electrocompetent E. coli cells ideal for construction of complex expression libraries and feature ≥1 x 108 cfu/µg efficiency with electroporation. BL21 (DE3) Electrocompetent E. coli cells have a widely used background, feature a T7 expression strain, are deficient in both lon (1) and ompT proteases. These cells are resistant to phage T1(fhuA2) and are B strain.